Astral™ Inoculation Device System

Economical, disposable inoculation system designed for safety and dependable sterility

Manufacturer: BIO PLAS INC 7000

 View more versions of this product

Catalog No. 05-664-80

  Check Availability
Add to Cart

    Specifications

    Disposable Yes
    For Use With (Equipment) Astral™ inoculation device system
    Includes Autoclavable handle
    Volume (Metric) 1μL
    Packaging Type Rack
    Sterile Yes
    Type Inoculating Loop
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit