Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand Autoclavable Pipet Basins
Eliminate the need for multiple reagent containerssave bench space.
Catalog Number | Type | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Type | Sterility | Price | Quantity & Availability | |||||
13-681-801
|
Pipette Basin | Non-sterile |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
13-681-800
|
Pipette Basin | Sterile |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
13-681-802
|
Multichannel Pipette Basin | Non-sterile |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Eliminate the need for multiple reagent containerssave bench space.
V-shaped basins facilitate easy withdrawal of sample solutions. Numbered chambers ensure accurate sample reference. Chemical-resistant polypropylene.
12-Channel Pipet Basins
- Designed for serial dilutions or pipetting individual reagents using up to a 12-channel pipetter
- Each chamber holds up to 5mL
- Withstand temperatures from -144° to +218°F (-80° to +121°C)
- Tight-fitting lid minimizes evaporation or contamination during incubation or storage
- Sterile or nonsterile; individually wrapped in easy-open bags
- Dimensions: 5L x 2.25W x 1.2 in.H (12.7 x 5.7 x 3.0cm)
- Unique design features three separate compartments: a 50mL single compartment with graduation marks at 10, 25, 40, and 50mL on one side; on the other side, an 8-channel compartment with eight 5mL reagent channels, and a 12-channel compartment with twelve 5mL reagent channels 5mL channels are numbered for easy reagent identification
- Use with all brands of multichannel pipetters
- Also ideal for filling multitube blocks and plates
- Dimensions: 4.75L x 4.3W x 1.1 in.H (12.1 x 10.9 x 2.8cm)
Specifications
Autoclavable | |
Polypropylene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title