Fisherbrand™ Autoclavable Pipet Basins

Eliminate the need for multiple reagent containers—save bench space

 View more versions of this product

Catalog No. 13-681-802

  Check Availability
Add to Cart

    Specifications

    Height (Metric) 2.8cm
    Material Polypropylene
    Autoclavable Yes
    Length (English) 4.75 in.
    Product Type Multichannel
    Length (Metric) 12.06cm
    Width (English) 4.3 in.
    Width (Metric) 10.9cm
    Dimensions (L x W x H) 4.75 x 4.3 x 1.1 in. (12.06 x 10.9 x 2.8cm)
    Sterile No
    Height (English) 1.1 in.

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit