Fisherbrand™ Automatic Burets with Stopcock

Ideal for storing standard solutions

 View more versions of this product

Catalog No. 03-842-1A

  Check Availability
Add to Cart

    Specifications

    Color Clear
    For Use With (Application) Ideal for storing standard solutions
    Includes Clear glass reservoir bottle, rubber stopper, one glass buret with central filling tube, rubber bulb, glass and rubber tubing connections, and glass stopcock
    Material Glass
    Capacity (Metric) 10mL
    Product Type Automatic Buret
    Subdivision 0.1mL

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit