Learn More
Fisherbrand™ General Protection Disposable SMS Assorted Colors Lab Coats
Single-use lab coats provide high durability and splash/dust protection
 View more versions of this product
Catalog No. 17-100-847
  Check Availability
Add to cart
Includes: 1 each of White, Blue, Pink, Cranberry; 2 each of Teal, Purple, Blueberry per pack
Specifications Description & Specifications
Specifications
Standard
Assorted
SMS (Spunbound Melt-Blown Synthetic)
5X-Large
Yes
General SMS protection; Three pockets
Snap
Knit
3
Yes
10/Pk.; 30/Cs.
50 in.