Fisherbrand™ General Protection Disposable SMS Assorted Colors Lab Coats
Single-use lab coats provide high durability and splash/dust protection
 View more versions of this product
Catalog No. 17-100-843
  Check Availability
Add to Cart
Includes: 1 each of White, Blue, Pink, Cranberry; 2 each of Teal, Purple, Blueberry per pack
    Specifications
    Disposable Yes
    Closure Type Snap
    Packaging Type 10/Pk.; 30/Cs.
    Product Type General SMS protection; Three pockets
    Length (English) 39 in.
    Material SMS (Spunbound Melt-Blown Synthetic)
    No. of Pockets 3
    Collar Type Standard
    Latex Free Yes
    Color Assorted
    Cuff Style Knit
    Size Large
  • Description & Specifications
Related Products
Product Content Feedback