Fisherbrand™ Basic Protection Polypropylene Disposable Lab Coats with Elastic Cuffs
Latex-free spunbond polypropylene construction lab ware
 View more versions of this product
Catalog No. 17-100-802
  Check Availability
Add to Cart
    Specifications
    Closure Type Snap
    Item Description 5-Snap
    No. of Pockets 0
    Collar Type Standard
    Latex Free Yes
    Cuff Style Elastic
    Material Polypropylene
    Size Medium
    Color White
  • Description & Specifications
Related Products
Product Content Feedback