Learn More
Fisherbrand™ Basic Protection Polypropylene Disposable Lab Coats with Elastic Cuffs
Latex-free spunbond polypropylene construction lab ware
 View more versions of this product
Catalog No. 17-100-807
  Check Availability
Add to cart
Specifications
Description
  • Single-use lab coat
  • Standard collar, no pockets
  • Packed 25 lab coats/case
  • Elastic cuffs hold sleeves in place
  • Five snaps for a secure closure
  • Fabric weight: 50g/m2
  • White
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Lab Coats with Pockets, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.
Description & Specifications
Specifications
Standard
Elastic
0
Yes
White
Polypropylene
5X-Large