Learn More
Fisherbrand™ Basic Protection SMS Disposable Lab Coats — Medical Blue
Catalog Number 17100874
Click to view available options
Size:
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Durable splash and dust protection
Single-use lab coats provide high durability and protection. Excellent particle retention and water vapor transmission (cool to wear).
  • Latex-free, three-layer SMS construction
  • Packed 30 lab coats/case
  • Available in a variety of colors
  • Knit cuffs hold sleeves in place; standard collar
  • Three pockets and a five-snap closure
  • Fabric weight: 50g/m2
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Teal, Purple, Cranberry, and Blueberry.
Size Medium
Color Medical Blue