Learn More
Fisherbrand™ Basic Protection SMS Disposable Lab Coats — Medical Blue
Click to view available options
Size:
Medium
Large
Description
Single-use lab coats provide high durability and protection. Excellent particle retention and water vapor transmission (cool to wear).
- Latex-free, three-layer SMS construction
- Packed 30 lab coats/case
- Available in a variety of colors
- Knit cuffs hold sleeves in place; standard collar
- Three pockets and a five-snap closure
- Fabric weight: 50g/m2
Specifications
Size | Medium |
Color | Medical Blue |