Learn More
3M™ Secure Click™ Respirator P95 Particulate Filter
Secure Click™ Hard Case D9000 Series Particulate Filters are approved for use with 3M™ Secure Click™ Respirators in environments containing dust, mists, and fumes.
Supplier: 3M™ D3071
Alert:
Not for use in environments that are immediately dangerous to life or health (IDLH), sandblasting, or in environments that contain asbestos.
Description
- Hard case recommended for harsh areas for easy decontamination and protection against sparks and high heat
- Filter case designed for certain spray applications
- Secure Click™ connection allows filters to be inserted for ease of assembly
- Solid top and channeled airflow help protect filter
- Common uses include filtering solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances
- May also be used for liquid or non-oil based particles from sprays that do not also emit harmful vapors and for metal fumes produced from welding, brazing, cutting and other operations involving heating of metals
Specifications
Particulate | |
Bayonet | |
Secure Click | |
Particulate Filter | |
95% | |
D7000 |
NIOSH Approved | |
White | |
P | |
No | |
Secure Click Half Facepiece HF-800 Reusable Respirators | |
100 Filters/Pk. |