Learn More
3M™ Secure Click™ Respirator P100 Particulate Filter
Secure Click™ Hard Case D9000 Series Particulate Filters are approved for use with 3M™ Secure Click™ Respirators in environments containing certain particles in grinding, sanding and welding applications.
Supplier: 3M™ D3091
Description
- Hard case recommended for harsh areas for easy decontamination and protection against sparks and high heat
- Filter case designed for certain spray applications
- Secure Click™ connection allows filters to be inserted for ease of assembly
- Solid top and channeled airflow help protect filter
- Swept-back design provides wide field of view
- For workplace/occupational use only
- Common uses of D3091 particulate filters include filtering solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances, as well as liquid or oil based particles from sprays that do not also emit harmful vapors, and for metal fumes produced from welding, brazing, cutting and other operations involving heating of metals
- May be used to help reduce exposure to radioactive particulate materials such as uranium and plutonium
- Can also be used for asbestos
Specifications
Particulate | |
Bayonet | |
Secure Click | |
Filter | |
100 | |
D3000 |
NIOSH Approved | |
Purple | |
P | |
No | |
Secure Click Half Facepiece HF-800 Reusable Respirators | |
100 Filters/Pk. |