Fisherbrand™ Biohazard Specimen Bags Available on GSA/VA Contract for Federal Government customers only.

Access pocket without opening sealed pouch

 View more versions of this product

Catalog No. 19-075-388D

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W) 10 x 6 in. (25 x 15cm)
    Length (English) 10 in.
    Length (Metric) 25cm
    Width (English) 6 in.
    Width (Metric) 15cm
    Product Type Biohazard Specimen Bag
    Certifications/Compliance FDA
    Includes Printed with biohazard graphics, zipper closure, 2mil thick pouch, 1.5mil thick pocket
    Material Polyethylene
    Thickness (Metric) 0.279mm
    For Use With (Application) Access pocket without opening sealed pouch
    Thickness (English) 2mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit