Fisherbrand™ Blood Collection Tray

Designed with phlebotomists in mind. Fisherbrand™ Blood Collection Tray was developed after extensive research in clinics and hospitals. This lightweight solution for organizing and transporting tubes and smaller items includes a sturdy plastic tray, 36-well tube rack and disposable inserts.

 View more versions of this product

Catalog No. 22-267-272

  Check Availability
Add to Cart

    Specifications

    Color White
    For Use With (Application) Blood collection
    For Use With (Equipment) 13mm tube rack and Blood Collection Tray (style A)
    Includes Extra-deep compartment
    Product Type Insert
    Material Polystyrene

    Designed specifically for blood collection supplies

    • Lightweight, roomy ABS tray includes a ratcheting handle with soft rubber grip that rotates out of the way for easy access
    • Disposable tray inserts keep your blood collection supplies clean and organized
    • Each tray includes a 36-place tube rack for either 13mm or 16mm test tubes
    • Trays feature a supply drawer and side pockets for extra storage space
    • Easy to clean: wash the tray and tube rack as needed and replace the insert periodically for a fresh start
    Disposable tray inserts
    • Each tray is shipped with two disposable inserts
    • Available in two configurations for use with either the 13mm or 16mm tube rack
    • Includes an extra-deep compartment that can be used as a sharps container
    • Additional inserts sold in packs of four

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit