Learn More
Fisherbrand™ Blue Epoxy-Coated Racks Products for Science Education
Heavy-gauge steel wire racks resist rust, corrosion, abrasives, organic solvents, acids and alkalis.
Specifications
Autoclavable Autoclavable
Color Blue
Material Epoxy-coated Heavy-gauge Steel Wire
Type Tube Rack
View More Specs
Products 1
Catalog Number Positions Holds Price Quantity & Availability  
Catalog Number Positions Holds Price Quantity & Availability  
S68122 Products for Science Education
View Documents
24 Tubes (30 to 40 mm) This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Specifications
Autoclavable
Blue
Epoxy-coated Heavy-gauge Steel Wire
Tube Rack