Fisherbrand™ Blue-Spirit™ Ever-Safe™ Laboratory Thermometers
Nontoxic and environmentally safe
 View more versions of this product
Catalog No. 15-160-25
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Immersion Length 76mm
    Temperature Range (English) 0° to 230°F
    • Permanently fused markings for long-term use
    • Contain a blue biodegradable liquid (isoamyl benzoate) and dye while providing accurate results
    All thermometers are available with PTFE coating for increased safety and to prevent breakage.
  • Description & Specifications
Related Products
Product Content Feedback