Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Borosilicate Glass Disposable Serological Pipets with Regular Tip, Short Length Available on GSA/VA Contract for Federal Government customers only.

Perfect for tissue culture work or procedures performed under fume hoods or in confined areas

 View more versions of this product

Catalog No. 13-678-36A


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.5mL
1mL
Borosilicate Glass
USP type I, ASTM E438
Yes
0.01mL
0.03 oz., 1mL
Individually Wrapped
0.03 oz.
Yes
Yellow
Serological Pipet
Regular
±0.04mL
Yellow
0.01mL
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit