Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bottle-Top Dispensers

Five fully adjustable models cover a dispensing range from 0.05 to 50mL


Includes: 38, 40 and 45mm adapters and an individual calibration certificate. Optional reservoirs are sold separately.


Products ${productFamilyLength}
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
03-692-180 Accessories Range: 1 to 30mL; Increments: 1mL N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-181 Accessories Range: 1 to 50mL; Increments: 1mL N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-179 Accessories Range: 0.2 to 10mL; Increments: 0.2mL N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-178 Accessories Range: 0.1 to 5mL; Increments: 0.1mL N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications

Description

Deliver even aggressive liquids and organic solvents safely and effortlessly (except hydrofluoric acid).

  • Precision valve mechanism ensures easy priming and minimum reagent waste, with no leakage back to the reservoir
  • Polypropylene protection sleeve surrounds the borosilicate glass barrel
  • Antidrip taps prevent accidental dispensing
  • Spring-lock provides quicker, more accurate volume selection
  • Dispenser head can be rotated 360° for convenient dispensing and volume adjustment

  • Smooth-moving PTFE piston minimizes operator fatigue and can be removed in seconds for cleaning
  • Dispensers fit directly onto 30mm-neck bottles
  • Accuracy: ±0.3% at maximum volume
  • Repeatability: ±0.1% or better
  • Only the liquid pathway is autoclavable to 121°C (250°F)

Certifications

CE marked
Manufactured to ISO 9001:2008 quality system standards

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit