Learn More
Fisherbrand™ Bourdon Vacuum Gauges Non-distribution product as customer accommodation.
Provide continual check on vacuum pumps and laboratory or industrial vacuum lines
Manufacturer:  Fisherbrand™ J4605
Catalog No. 11-279A
Add to cart
Description
Description
Drawn steel case with twist-on crystal (2.5 in.) or ring and glass (4.5 in.), enamel finish, polycarbonate movement.
  • Phosphor bronze Bourdon tube reads vacuum in 0 to 30 in.Hg (101kPa) range
  • Accuracy is ±2% in midrange, ±3% at extreme
  • Vacuum connection of the 4.5 in. diameter gauge has T-handle gauge cock with 0.25 NPT female connection
Order Info
T-handle gauge cock must be purchased separately for the 2.5 in. gauge; contact your Fisher Scientific Customer Service Representative to order.