Fisherbrand™ Calcium Alginate Swabs
Soft calcium alginate swabs contain no fatty acids that can inhibit the growth of many fastidious microorganisms
 View more versions of this product
Catalog No. 14-959-82
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
  • Description & Specifications
Related Products
Product Content Feedback