Learn More
Fisherbrand™ Cervex-Brush™ Combination
Delivers higher yield of endocervical cells
Catalog No. 14-560-100
Add to cart
Description
Description
  • Single-collection device combines the features of the Cervex-Brush (14-372-53) and EndoCervex™ Brush-S (14-560-101)
  • Longer middle hairs of the EndoCervex have been replaced by a complete endocervical sampler
  • Maximizes simultaneous collection of ectocervical and endocervical cells in all patients
  • Requires only two full clockwise turns
  • Enhances specimen-to-slide transfer
Specifications
Specifications
Delivers higher yield of endocervical cells, enhances specimen-to-slide transfer
50 Pack
Cervex-Brush Combination