Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Class 100 Cleanroom Laminated Top Shelf with Built-In LED Light

Built-in undershelf LED light provides extra light on the work surface for a specific work or detail that requires higher brightness than existing light fixture. LED light does not have toxic mercury gas and buzzing sound as in fluorescent bulb.

$251.50 - $329.00

Specifications

Depth (English) Shelf 12 in.
Color White
Type Cleanroom Laminated Top Shelf with Built-In LED Light
View More Specs

Includes: One piece laminated top shelf, one pair of shelf bracket and one piece of power adapter

Products 3
Catalog Number Width (English) Shelf Price Quantity & Availability  
Catalog Number Width (English) Shelf Price Quantity & Availability  
25-100-363
View Documents
48 in.
Each for $251.50
Add to cart
 
25-100-364
View Documents
60 in.
Each for $291.50
Add to cart
 
25-100-365
View Documents
72 in.
Each for $329.00
Add to cart
 
Description

Description

  • Class 100 Cleanroom and anti static
  • Height adjustable top shelf
  • Plug and play, no-tool-assembly
  • Built-in LED touch-sensitive light with dimming feature Light Power Input: 100 -240V, 50/60Hz; Output: 12V DC
  • 100 lb. capacity per shelf

Warranty and Services

25 year unconditional manufacturer warranty

Specifications

Specifications

12 in.
White
Cleanroom Laminated Top Shelf with Built-In LED Light
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit