Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Class B Amber Glass Threaded Vials Available on GSA/VA Contract for Federal Government customers only.

General-purpose vials for light-sensitive materials

 View more versions of this product

Catalog No. 03-339-23D



Add to cart

Includes: Black phenolic screw-top closures with white rubber liners

Description

Description

General-purpose vials for light-sensitive materials

  • Made from Type I, Class B amber borosilicate glass
  • Autoclavable
  • Black screw-top cap
  • White rubber liner
  • Caps packed separately

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Closures are packaged separately

Specifications

Specifications

Threaded Vial
Amber
Borosilicate Glass
16mL
B
18-400
Yes
70mm
Rubber
4.32 dr.
21mm
Non-sterile
Yes
Phenolic
Screw cap
For general purpose vials for light sensitive materials
70mm
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit