Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Class B Clear Glass Threaded Vials

General purpose vial avoids alteration in pH of sample

 View more versions of this product

Catalog No. 03-339-22N



Add to cart

Includes: Black phenolic screw-top closures with white rubber liners

Description

Description

General purpose vial avoids alteration in pH of sample

  • Made from Type I, Class B borosilicate glass
  • Autoclavable

Order Info

Closures are packaged separately

Specifications

Specifications

Yes
Phenolic
Screw cap
For general purpose
Rubber
11 dr.
28 mm
Non-sterile
B
24-400
Yes
108mm
Sample Vial
Clear
Borosilicate Glass
41 mL
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit