Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Class B Clear Glass Threaded Vials with Closures Attached

General-purpose vial

 View more versions of this product

Catalog No. 03-338J



Add to cart

Includes: Attached black phenolic screw-top closures with polyvinyl-faced pulp liner

Description

Description

General-purpose vial

  • Made from Type I, Class B borosilicate glass
  • Attached black phenolic screw-top closures with polyvinyl-faced pulp liner
  • GPI thread finish

Specifications

Specifications

Sample Vial
Clear
Borosilicate Glass
29.6mL
Polyvinyl
Phenolic
Screw cap
For general purpose
8 dr.
25mm
Non-sterile
95mm
B
22-400
Yes
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit