Learn More
Fisherbrand™ Class B Cylinders with Single Amber Metric Scale Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Deliver”
 View more versions of this product
Catalog No. 08-570A
Add to cart
Description
Description
Calibrated “To Deliver”
  • Ideal for use with light-colored liquids
  • Dark amber graduations provide contrast for easy readings
  • Glass cylinders with heavy round bases and pouring spouts
  • Accuracy is assured even at lowest graduations; first 10% of volume is not graduated
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
Designed from ASTM™ E 1272, Style 1, Class B requirements.
Specifications
Specifications
120mm
ASTM E1272
5mL
4.7 in.
±0.1mL
Yes
0.1mL
Borosilicate Glass
B
Ideal for use with light colored liquids, amber graduations provide contrast for easy readings, heavy round bases and pouring spouts
Single amber metric scale
0.1mL
+/- 0.10mL