Learn More
Fisherbrand™ Class B Mixing Cylinders Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain”
 View more versions of this product
Catalog No. 08-565D
Add to cart
Description
Description
Calibrated “To Contain”
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Borosilicate Glass
100mL
Pennyhead
Glass
Yes
1mL
B
Double white enamel metric graduations
±0.6mL
1mL
+/- 0.6mL