Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Classic Glass Thistle Tube Funnels

Funnels manufactured from borosilicate glass with a thistle top.
Description
- Top diameter: 45mm
- Stem diameter: 6mm
Specifications
45mm | |
Borosilicate Glass | |
Thistle Tube Funnel |
6mm | |
250mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title