Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cleaning Solution (Chromic-Sulfuric Acid) Available on GSA/VA Contract for Federal Government customers only.

Removes nearly impossible stains from glassware

Specifications

Boiling Point 290°C
Color Dark Brown
Format Liquid
View More Specs

Products 1
Catalog Number Size Price Quantity & Availability  
Catalog Number Size Price Quantity & Availability  
SC88-500 Available on GSA/VA Contract for Federal Government customers only.
View Documents
500mL
Each for $75.00
Case of 6 EA for $361.00
Order in Bulk

Request bulk or custom formats

N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Premixed

Order Info

Available with tough Safe-Cote™ plastic to help prevent leakage or flying glass should bottle accidentally break

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

290°C
Dark Brown
Liquid
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit