Learn More
Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap Available on GSA/VA Contract for Federal Government customers only.
For storing liquid samples. Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap are clear for easy viewing of contents.
$129.03
Specifications
Closure Liner Poly/Vinyl
Closure Type Screw
Closure Size 22-400
Color Clear, Clear
Material Glass, Glass
View More Specs
Products 5
Catalog Number Capacity (Metric) Closure Size Quantity Price Quantity & Availability  
Catalog Number Capacity (Metric) Closure Size Quantity Price Quantity & Availability  
02-911-778 View Documents 960mL 33-400 12/Cs. Case of 12 for $129.03
Add to cart
02-911-780 Available on GSA/VA Contract for Federal Government customers only. View Documents 120mL 22-400 24/Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-911-777 View Documents 480mL 28-400 12/Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-911-752 View Documents 60mL 20-400 24/Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-911-781 View Documents 240mL 24-400 12/Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Clear Glass Boston Round Bottles are excellent general use bottles ideal for when content integrity and sample or product visibility is important. Boston Round bottles feature a narrow screw neck opening, curved shoulders, and a round body. Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary. These environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage.
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Clear flint glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Poly/Vinyl
22-400
Glass, Glass
Screw
Clear, Clear
Bottle, Boston Round, Boston Round Bottle