Learn More
Fisherbrand™ Clear French Square Bottles with White Polypropylene Pulp/Vinyl Cap
Space-saving and easy to fill. Fisherbrand™ Clear French Square Bottles with White Polypropylene Pulp/Vinyl Lined Cap features a wide mouth opening ideal for mixing, storing and sampling.
 View more versions of this product
Catalog No. 02-912-017
Add to cart
Alert:
Not recommended for active hydrocarbons or bleaches.
Description
Description
Clear (Flint) Glass French Square Bottles are space-saving and easy to fill. The clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents. The French Square bottle design allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space. The wide mouth opening is ideal for mixing, storing and sampling.
Clear French Square Bottles
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle contents
  • French Square design allows bottle to hold maximum volume of contents while taking up minimum shelf and storage space
Polypropylene Pulp/Vinyl Lined Cap
  • Offer good impact strength and pliability
  • Can be remelted after they are formed
  • Vinyl coating is applied to high density polyethylene coated paper and then laminated
Specifications
Specifications
32 oz., 1000mL
32oz.
Pulp/Vinyl
Yes
12/Cs.
White
Pulp/Vinyl
58-400
Screw
960mL
Polypropylene
Clear, Clear
Glass, Glass
Bottle, Graduated French Square, French Square Bottle
58-400
Wide
French Square