Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear French Square Bottles with White Polypropylene Pulp/Vinyl Cap

Space-saving and easy to fill. Fisherbrand™ Clear French Square Bottles with White Polypropylene Pulp/Vinyl Lined Cap features a wide mouth opening ideal for mixing, storing and sampling.

 View more versions of this product

Catalog No. 02-912-302


  Check Availability
Add to cart

Alert:

Not recommended for active hydrocarbons or bleaches.


Description

Description

Clear (Flint) Glass French Square Bottles are space-saving and easy to fill. The clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents. The French Square bottle design allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space. The wide mouth opening is ideal for mixing, storing and sampling.

Clear French Square Bottles

  • Clear glass provides maximum content visibility and minimal chemical inference with bottle contents
  • French Square design allows bottle to hold maximum volume of contents while taking up minimum shelf and storage space
Polypropylene Pulp/Vinyl Lined Cap
  • Offer good impact strength and pliability
  • Can be remelted after they are formed
  • Vinyl coating is applied to high density polyethylene coated paper and then laminated

Specifications

Specifications

0.5 oz., 15mL
0.5 oz.
Pulp/Vinyl
GlassþGlass
Bottle, French SquareþFrench Square Bottle
20-400
Wide
French Square
15mL
Polypropylene
ClearþClear
576/Cs.
White
Pulp/Vinyl
20-400
Screw
Documents
Promotions

Promotions

  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.

    06-30-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit