Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear French Square Bottles with Black Phenolic Rubber Cap

Ideal for mixing, storing and sampling. Fisherbrand™ Clear French Square Bottles with Black Phenolic Rubber Lined Cap are space-saving and easy to fill.

 View more versions of this product

Catalog No. 02-911-396


  Check Availability
Add to cart

Description

Description

Clear (Flint) Glass French Square Bottles are space-saving and easy to fill. The clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents. The French Square bottle design allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space. The wide mouth opening is ideal for mixing, storing and sampling.

  • Clear glass provides maximum content visibility and minimal chemical inference with bottle contents
  • Allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space

Specifications

Specifications

16 oz., 500mL
16 oz.
ClearþClear
24/Cs.
Black
Rubber
48-400
Screw
480mL
Phenolic
GlassþGlass
Bottle, French SquareþFrench Square Bottle
48-400
Wide
French Square
Documents
Promotions

Promotions

  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.

    06-30-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit