Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear Glass Jugs with Black Phenolic Rubber Cap Available on GSA/VA Contract for Federal Government customers only.

Fisherbrand™ Clear Glass Jugs with Black Phenolic Rubber Cap are for general use.

 View more versions of this product

Catalog No. 02-912-275


  Check Availability
Add to cart

Description

Description

Clear / Flint Glass Jugs with molded finger grips are the perfect storage solution for large quantities of liquid.

  • Type III soda-lime glass
  • Narrow mouth enables careful pouring and prevents spillage
  • 38mm-400 screw caps attached
  • With molded finger grips

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

65 oz., 1920mL
65 oz.
Rubber
GlassþGlass
Jug, RoundþJug
38-400
Narrow
Round
1920mL
Phenolic
ClearþClear
6/Cs.
Black
Rubber
38-400
Screw
Documents
Promotions

Promotions

  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.

    06-30-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit