Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear Glass Jugs

Catalog No. 02-912-272


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

130 oz., 3844.56mL
128 oz.
Polyethylene
Glass
Jug, Round
38-400
Narrow
Round
3785mL
Phenolic
Clear
4/Cs.
Black
PolyCone
38-400
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit