Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear Square Tablet Bottles with Black Phenolic PolyCone Cap Available on GSA/VA Contract for Federal Government customers only.

Provides maximum content visibility. Fisherbrand™ Clear Square Tablet Bottles with Black Phenolic PolyCone Cap is available in capacities of 1 and 2 oz. and screw cap size of 33mm-400.

 View more versions of this product

Catalog No. 02-911-993


  Check Availability
Add to cart

Description

Description

Clear / Flint Square Tablet Bottles provide maximum content visibility. Square glass bottles feature a space saving design for bench tops with limited storing space. These wide mouth bottles are ideal for mixing, storing and sampling.

  • Feature a space saving design for bench tops with limited storing space
  • Wide mouth bottles are ideal for mixing, storing and sampling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

1 oz., 30mL
1 oz.
Polyethylene
GlassþGlass
Bottle, Square TabletþSquare Tablet Bottle
33-400
Wide
Square
30mL
Phenolic
ClearþClear
288/Cs.
Black
PolyCone
33-400
Screw
Documents
Promotions

Promotions

  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.

    06-30-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit