Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear PET Packer Bottles with White Polypropylene PE Foam Cap Available on GSA/VA Contract for Federal Government customers only.

Ideal for oil analysis. Fisherbrand™ Clear PET Packer Bottles with White Polypropylene PE Foam Cap provides a good alcohol and solvent barrier.

 View more versions of this product

Catalog No. 02-911-808


  Check Availability
Add to cart

Description

Description

Clear PET Packer Bottles provide a good alcohol and solvent barrier. These clear and transparent bottles make sample identification a snap. PET is stress and impact resistant. Ideal for oil analysis.

  • Clear and transparent for easy sample identification
  • PET is stress and impact resistant
  • Provides a good alcohol and solvent barrier

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

8.45 oz., 250mL
8.5oz.
Polypropylene
ClearþClear
48/Cs.
White
PE Foam
45-400
Screw
250mL
ISO 9001
Foam Polyethylene
PETþPET
Bottle, Wide Mouth PackerþPacker Bottle
45-400
Wide
Round
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit