Learn More
Fisherbrand™ Clear PVC Tubing Available on GSA/VA Contract for Federal Government customers only.
General-purpose polyvinyl chloride tubing for rinse and drain tubes
 View more versions of this product
Catalog No. 14-169-7B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PVC
0.188 in.
4.8mm
50 ft.
1.6mm
0.3125 in.
0.0625 in.
7.9mm
15.2m