Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap Available on GSA/VA Contract for Federal Government customers only.

For storing dry materials and specimens. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap is an ideal general purpose bottle

 View more versions of this product

Catalog No. 02-911-916

  Check Availability
Add to Cart

    Specifications

    Product Type Standard Wide Mouth Bottle
    Closure Color White
    Closure Type Screw
    Liner Material Pulp/Vinyl
    Mouth Wide
    Neck Finish 58-400
    Quantity 24/Cs.
    Shape Round
    Capacity (Metric) 240mL
    Capacity (English) 8oz.
    Closure Material Polypropylene
    Material Glass
    Closure Screw
    Color Clear
    Closure Liner Poly/Vinyl
    Closure Size 58-400
    Capacity 8 oz., 250mL

    • Features a wide mouth opening for easy transfer of contents
    • Applications include pharmaceutical, chemical, environmental, and food packaging and storing

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit