Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear Straight-Sided Jars with Polyvinyl-Lined Cap Available on GSA/VA Contract for Federal Government customers only.

For easy viewing of contents. Fisherbrand™ Clear Straight-Sided Jars with Polyvinyl-Lined Cap are for multipurpose use.

 View more versions of this product

Catalog No. 02-911-775

  Check Availability
Add to cart

Specifications

Description

  • Designed without shoulders for maximum storage capacity
  • Available in a wide variety of sizes, from 1 oz. to 32 oz.
Description & Specifications

Specifications

4 oz., 125mL
4 oz.
Poly/Vinyl
Glass
Jar, Straight Sided
58-400
Wide
Straight Sided Jar
120mL
Polypropylene
Clear
24/Cs.
White
Pulp/Vinyl
58-400
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit