Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clear Straight-Sided Jars with PTFE-Faced PE-Lined Cap

Clear multipurpose jars for maximum content visibility. Qorpak™ Clear Straight-Sided Jars with PTFE-Faced PE-Lined Cap are available in in a wide variety of sizes, from 1 to 32 oz.

 View more versions of this product

Catalog No. 02-911-762


  Check Availability
Add to cart

Specifications

Description

  • Designed without shoulders for maximum storage capacity
  • Wide mouth for easy filling
Description & Specifications

Specifications

32 oz., 1000mL
32oz.
Polyethylene
Glass
12/Cs.
White
PE Foam/PTFE
89-400
Screw
960mL
Polypropylene
Clear
12/Cs.
Jar, Straight Sided
89-400
Wide
Straight Sided Jar
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit