Learn More
Fisherbrand™ Color-Coded Dumbbell-Style Stir Bars
GSA_VA
Provide fast mixing action, strong turbulence, greater stability and less drifting at higher speeds.
$27.00 - $30.20
Specifications
Quantity 50
Shape Dumbbell
View More Specs
Products 4
Catalog Number Color Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Color Length (English) Length (Metric) Price Quantity & Availability  
14-512-158
GSA_VA
View Documents
Red 1.25 in. 36mm
Each for $27.00
Case of 50 EA for $1,040.00
Add to cart
14-512-159
View Documents
Blue 1.25 in. 36mm
Each for $28.10
Case of 50 EA for $1,088.00
Add to cart
14-512-160
GSA_VA
View Documents
Red 2 in. 54mm
Each for $30.20
Case of 50 EA for $1,190.00
Add to cart
14-512-161
GSA_VA
View Documents
Blue 2 in. 50.8mm
Each for $30.20
Case of 50 EA for $1,189.00
Add to cart
Description
Description
  • Ideal for containers with round or convex bottoms
  • All sizes have solid FDA grade, USP Class VI Teflon™ PTFE end disks 20mm (0.79 in.) in diameter, a bar diameter of 8mm (0.315 in.), and an Alnico magnet
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
50
Dumbbell