Learn More
Fisherbrand™ Color-Coded TainerTop™ Safety Closures Available on GSA/VA Contract for Federal Government customers only.
Seven colors correspond to Vacutainer™ cap colors, ensuring easy identification
$121.00 - $127.00
Specifications
Autoclavable No
Material Polyethylene
For Use With (Application) Resealing glass test tubes and glass culture tubes
View More Specs
Products 27
Catalog Number Color For Use With (Equipment) Price Quantity & Availability  
Catalog Number Color For Use With (Equipment) Price Quantity & Availability  
14-376-61 View Documents Red 10mm culture tube Pack of 1000 for $127.00
Case of 10 PK for $1,095.99
Add to cart
14-376-60 View Documents White 10mm culture tube Pack of 1000 for $127.00
Case of 10 PK for $1,095.99
Add to cart
14-376-62 View Documents Lavender 10mm culture tube Pack of 1000 for $127.00
Case of 10 PK for $1,095.99
Add to cart
14-376-63 View Documents Green 10mm culture tube Pack of 1000 for $127.00
Case of 10 PK for $1,095.99
Add to cart
14-376-69 Available on GSA/VA Contract for Federal Government customers only. View Documents White 13mm culture tubes Pack of 1000 for $121.00
Case of 10 PK for $1,051.85
Add to cart
14-376-64 View Documents Blue 10mm culture tube Pack of 1000 for $127.00
Case of 10 PK for $1,095.99
Add to cart
14-376-67 View Documents Gray 13mm culture tubes Pack of 1000 for $122.00
Case of 10 PK for $1,055.15
Add to cart
14-376-66 View Documents Yellow 10mm culture tube Pack of 1000 for $127.00
Case of 10 PK for $1,095.99
Add to cart
14-376-70 Available on GSA/VA Contract for Federal Government customers only. View Documents Red 13mm culture tubes Pack of 1000 for $121.00
Case of 10 PK for $1,051.85
Add to cart
14-376-71 Available on GSA/VA Contract for Federal Government customers only. View Documents Lavender 13mm culture tubes Pack of 1000 for $121.00
Case of 10 PK for $1,051.85
Add to cart
14-376-75 Available on GSA/VA Contract for Federal Government customers only. View Documents Red 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-72 Available on GSA/VA Contract for Federal Government customers only. View Documents Green 13mm culture tubes Pack of 1000 for $121.00
Case of 10 PK for $1,051.85
Add to cart
14-376-74 Available on GSA/VA Contract for Federal Government customers only. View Documents White 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-73 Available on GSA/VA Contract for Federal Government customers only. View Documents Blue 13mm culture tubes Pack of 1000 for $121.00
Case of 10 PK for $1,051.85
Add to cart
14-376-76 Available on GSA/VA Contract for Federal Government customers only. View Documents Lavender 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-77 Available on GSA/VA Contract for Federal Government customers only. View Documents Green 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-78 Available on GSA/VA Contract for Federal Government customers only. View Documents Blue 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-79 View Documents Gray 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-82 Available on GSA/VA Contract for Federal Government customers only. View Documents Yellow 16mm culture tube Pack of 1000 for $122.00
Case of 10 PK for $1,046.03
Add to cart
14-376-80 Available on GSA/VA Contract for Federal Government customers only. View Documents Yellow 12mm culture tube and 13mm blood collection tube Pack of 1000 for $122.00
Case of 10 PK for $1,057.93
Add to cart
14-376-83 Available on GSA/VA Contract for Federal Government customers only. View Documents White 16mm culture tube Pack of 1000 for $124.00
Case of 10 PK for $1,065.28
Add to cart
14-376-81 View Documents Gray 16mm culture tube Pack of 1000 for $122.00
Case of 10 PK for $1,064.42
Add to cart
14-376-85 Available on GSA/VA Contract for Federal Government customers only. View Documents Lavender 16mm culture tube Pack of 1000 for $122.00
Case of 10 PK for $1,048.56
Add to cart
14-376-86 Available on GSA/VA Contract for Federal Government customers only. View Documents Green 16mm culture tube Pack of 1000 for $122.00
Case of 10 PK for $1,046.87
Add to cart
14-376-84 Available on GSA/VA Contract for Federal Government customers only. View Documents Red 16mm culture tube Pack of 1000 for $124.00
Case of 10 PK for $1,065.28
Add to cart
14-376-87 Available on GSA/VA Contract for Federal Government customers only. View Documents Blue 16mm culture tube Pack of 1000 for $122.00
Case of 10 PK for $1,044.36
Add to cart
14-376-88 Available on GSA/VA Contract for Federal Government customers only. View Documents Yellow 13mm culture tubes Pack of 1000 for $121.00
Case of 10 PK for $1,051.85
Add to cart
Description
Description
Seven colors correspond to Vacutainer™ cap colors, ensuring easy identification
Recap blood drawing tubes, disposable glass test tubes, and plastic culture tubes. Protect from aerosols of highly infectious organisms such as TB and the HIV virus. Guard samples against cross-contamination, and lab work areas against infectious agents and mess.
  • Polyethylene
  • Information can be written directly on cap with indelible ink
  • Ensure integrity of serum samples after freezing or en route to reference labs
  • No precutting required
  • Won't pop or crack when frozen
  • Provide liquid-tight seal — will not aerosol when opened
  • Hold fast under most rigorous procedures including agitation, centrifugation, refrigeration, and freezing
  • Flexible closures are not autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
No
Polyethylene
Resealing glass test tubes and glass culture tubes