Learn More
Fisherbrand™ Color-Coded TainerTop™ Safety Closures Available on GSA/VA Contract for Federal Government customers only.
Seven colors correspond to Vacutainer™ cap colors, ensuring easy identification
 View more versions of this product
Catalog No. 14-376-73
Add to cart
Description
Description
Seven colors correspond to Vacutainer™ cap colors, ensuring easy identification
Recap blood drawing tubes, disposable glass test tubes, and plastic culture tubes. Protect from aerosols of highly infectious organisms such as TB and the HIV virus. Guard samples against cross-contamination, and lab work areas against infectious agents and mess.
  • Polyethylene
  • Information can be written directly on cap with indelible ink
  • Ensure integrity of serum samples after freezing or en route to reference labs
  • No precutting required
  • Won't pop or crack when frozen
  • Provide liquid-tight seal — will not aerosol when opened
  • Hold fast under most rigorous procedures including agitation, centrifugation, refrigeration, and freezing
  • Flexible closures are not autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
No
Blue
13mm culture tubes
Polyethylene
Resealing glass test tubes and glass culture tubes