Learn More
Fisherbrand™ Color-Coded Biohazard Specimen Transport Bags
Color-coded bag lip for easy identification
 View more versions of this product
Catalog No. 01-800-13
Add to cart
Description
Description
  • Measure 6 x 9 in. (15 x 23cm)
  • Printed with orange biohazard symbol, blue stat and storage instructions
  • Document pouch keeps paperwork separate from specimens
Order Info
Packaged 100 per pack, 10 packs per case; assorted case contains 200 bags of each color
Specifications
Specifications
Zip closure
9 in.
Red
PE (Polyethylene)
9 x 6 in. (23 x 15cm)
6 in.
23cm
15cm
Documents
Accessories