Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Colored Label Tape: Rainbow Pack Get IT Item Available on GSA/VA Contract for Federal Government customers only.

Oil resistant, waterproof, acid resistant tape excellent for color coding.

 View more versions of this product

Catalog No. 15-901-5R

  Check Availability
Add to cart

Specifications

Description

Available on 14 yd. (13m) rolls.

  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily

Tapes are supplied without dispenser.

Description & Specifications

Specifications

Multi-colored
Oil resistant, Waterproof, Acid resistant and excellent for color coding
14 yd.
-10 to 250°F
0.75 in.
Colored Label Tape
19mm x 13m
2 Blue, 2 pink, 2 green, 2 red, 2 yellow, 2 orange, 4 white
13m
-23 to 121°C
Tape
16 Case
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit