Learn More
Fisherbrand™ ColorFrost™ Disposable Microscope Slides Available on GSA/VA Contract for Federal Government customers only.
Color-coded frosted end for easy coordinating with tissue cassettes or other color systems
 View more versions of this product
Catalog No. 22-214361
Add to cart
Description
Description
Color-coded frosted end for easy coordinating with tissue cassettes or other color systems
  • Plain corners
  • Same properties as Superfrost™, including chemical resistance and easy marking and reading
  • Convenient “SPECIMEN” mark lends easy orientation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Thickness: 1.0mm.
Specifications
Specifications
Chemical Resistant
Plain
Colorfrost
Microscope Slide
75mm
Yellow
76 x 25mm
25mm
1mm