Learn More
Fisherbrand™ ColorFrost™ Disposable Microscope Slides
Color-coded frosted end for easy coordinating with tissue cassettes or other color systems
$126.00 - $144.90
Specifications
Frosted Colorfrost
Thickness (Metric) 1 mm
Type Microscope Slide
Coating Chemical Resistant
Corner Style Plain
View More Specs
Products 13
Catalog Number Color Dimensions (L x W) Price Quantity & Availability  
Catalog Number Color Dimensions (L x W) Price Quantity & Availability  
22-037077
View Documents
Aqua 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
22-037079
View Documents
Burnt Orange 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
22-037080
View Documents
Gold 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
22-037078
View Documents
Tan 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
22-037081
View Documents
Lilac 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
22-214320
View Documents
Blue 76 x 25 mm
Pack of 144 for $144.90
Case of 10 PK for $1,235.00
Add to Cart
22-214346
View Documents
Pink 76 x 25 mm
Pack of 144 for $144.90
Case of 10 PK for $1,235.00
Add to Cart
22-214361
View Documents
Yellow 76 x 25 mm
Pack of 144 for $144.90
Case of 10 PK for $1,235.00
Add to Cart
22-214338
View Documents
Green 76 x 25 mm
Pack of 144 for $144.90
Case of 10 PK for $1,235.00
Edge
Add to Cart
12-550-413
View Documents
Green 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
12-550-403
View Documents
Blue 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
12-550-423
View Documents
Pink 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
12-550-433
View Documents
Yellow 75 x 25 mm
Pack of 144 for $126.00
Case of 10 PK for $1,050.00
Add to Cart
Description
Description
Color-coded frosted end for easy coordinating with tissue cassettes or other color systems
  • Plain corners
  • Same properties as Superfrost™, including chemical resistance and easy marking and reading
  • Convenient “SPECIMEN” mark lends easy orientation
Order Info
Thickness: 1.0mm.
Specifications
Specifications
Colorfrost
Microscope Slide
Plain
1 mm
Chemical Resistant