Fisherbrand™ Cotton Canvas Gloves

Protection with comfort

 View more versions of this product

Catalog No. 19-013-526

  Check Availability
Add to Cart

    Specifications

    Weight (Metric) 227g
    Cut Style Clute
    Glove Cuff Style Knit
    Grade Economy
    For Use With (Application) multipurpose Fabric gloves used for light protection in many general applications
    Material Cotton, Canvas
    Powdered Yes
    Size Women's
    Sterile Yes
    Weight (English) 8 oz.
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit