Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protective Industries Cotton Canvas Gloves

Protection with comfort

Manufacturer:  Protective Industries 90912

 View more versions of this product

Catalog No. 19-120-1652



Add to cart

Description

Description

Multi-purpose fabric gloves used for light protection in many general applications.

  • Heavier weights provide improved protection
  • Blended cotton/polyester fabric (except 12-oz. size, which is 100% cotton)
  • Knitwrist; Clute cut
  • Color: Natural

Specifications

Specifications

Clute
Knit
light protection in many general applications, including construction, material handling and general maintenance
Men's
340.2g
5.75 in.
Natural
26.67cm
10.5 x 5.75 in. (26.67 x 14.5cm)
Premium
Cotton
Heavy weight
12 oz.
14.5cm
10.5 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit